(2S)-2-(4-Bromophenyl)-2-hydroxyacetic acid ≥ ee 99%
Dodavatel: Apollo Scientific
OR310335-1G
OR310335-500MG
APOSOR310335-1GEA
30290
CZK
APOSOR310335-1G
APOSOR310335-500MG
(2S)-2-(4-Bromophenyl)-2-hydroxyacetic acid ≥ ee 99%
(2S)-2-(4-Bromophenyl)-2-hydroxyacetic acid
Vzorec:
C₈H₇BrO₃ MW: 231.043 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798440 CAS číslo: 123484-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...