5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-carboxamide
OR310332-1G
OR310332-500MG
APOSOR310332-1GEA
19110
CZK
APOSOR310332-1G
APOSOR310332-500MG
5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide
5-Amino-1-(oxan-4-yl)-1H-pyrazole-4-carboxamide
Vzorec:
C₉H₁₄N₄O₂ MW: 210,233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12405839 CAS číslo: 1082745-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...