Tisk…
Ethyl 1-(5-formylpyridin-2-yl)-1H-pyrazole-4-carboxylate
Dodavatel: Apollo Scientific
OR310322-1G OR310322-500MG
APOSOR310322-1GEA 25460 CZK
APOSOR310322-1G APOSOR310322-500MG
Ethyl 1-(5-formylpyridin-2-yl)-1H-pyrazole-4-carboxylate
Ethyl 1-(5-formylpyridin-2-yl)-1H-pyrazole-4-carboxylate
Vzorec: C₁₂H₁₁N₃O₃
MW: 245.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23579693
CAS číslo: 1505914-27-1