6-Methylbenzo[b]thiophene-2-carboxylic acid 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Carboxy-6-methyl-1-benzothiophene, 6-Methyl-1-benzothiophene-2-carboxylic acid
OR3103-1G
OR3103-5G
OR3103-25G
APOSOR3103-1GEA
622
CZK
APOSOR3103-1G
APOSOR3103-5G
APOSOR3103-25G
6-Methylbenzo[b]thiophene-2-carboxylic acid 99%
6-Methylbenzo[b]thiophene-2-carboxylic acid
Vzorec:
C₁₀H₈O₂S MW: 192,234 g/mol Bod tání: 243…244 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07377056 CAS číslo: 1467-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...