2-Bromo-1-methylnaphthalene 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR310251-1G
OR310251-250MG
APOSOR310251-1GEA
0
CZK
APOSOR310251-1G
APOSOR310251-250MG
2-Bromo-1-methylnaphthalene 95%
2-Bromo-1-methylnaphthalene
Vzorec:
C₁₁H₉Br MW: 221,093 g/mol Bod tání: 31…37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10000779 CAS číslo: 20601-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...