8-Chloro-2-oxo-2H-chromene-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR31023-1G
OR31023-250MG
APOSOR31023-1GEA
4270
CZK
APOSOR31023-1G
APOSOR31023-250MG
8-Chloro-2-oxo-2H-chromene-3-carboxylic acid
8-Chloro-2-oxo-2H-chromene-3-carboxylic acid
Vzorec:
C₁₀H₅ClO₄ MW: 224,597 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13187195 CAS číslo: 107747-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...