Tisk…

2-Chloro-5-iodo-3-methylpyridine

Dodavatel: Apollo Scientific

OR310157-5G OR310157-25G
APOSOR310157-5GEA 2650 CZK
APOSOR310157-5G APOSOR310157-25G
2-Chloro-5-iodo-3-methylpyridine
2-Chloro-5-iodo-3-methylpyridine
Vzorec: C₆H₅ClIN
MW: 253.468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037455
CAS číslo: 59782-89-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR