6-Methoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
OR31014-1G
OR31014-250MG
APOSOR31014-1GEA
32730
CZK
APOSOR31014-1G
APOSOR31014-250MG
6-Methoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
6-Methoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Vzorec:
C₁₃H₁₆N₂O₃·ClH MW: 284,739 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22372630 CAS číslo: 1204346-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...