Tisk…

2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole

Dodavatel: Apollo Scientific

OR310136-1G OR310136-500MG
APOSOR310136-1GEA 19110 CZK
APOSOR310136-1G APOSOR310136-500MG
2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole
2-Bromo-5-(4-nitrophenoxy)-1,3,4-thiadiazole
Vzorec: C₈H₄BrN₃O₃S
MW: 302,105 g/mol
Bod tání: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD20368360
CAS číslo: 1343454-17-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR