Ethyl 2-ethoxy-4-oxo-4,5-dihydrofuran-3-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR31013-1G
OR31013-250MG
APOSOR31013-1GEA
12200
CZK
APOSOR31013-1G
APOSOR31013-250MG
Ethyl 2-ethoxy-4-oxo-4,5-dihydrofuran-3-carboxylate
Ethyl 2-ethoxy-4-oxo-4,5-dihydrofuran-3-carboxylate
Vzorec:
C₉H₁₂O₅ MW: 200,189 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00667747 CAS číslo: 36717-48-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...