Tisk…

4-(Benzylamino)furan-2(5H)-one

Dodavatel: Apollo Scientific
Varování

OR31011-1G OR31011-250MG
APOSOR31011-1GEA 23150 CZK
APOSOR31011-1G APOSOR31011-250MG
4-(Benzylamino)furan-2(5H)-one
4-(Benzylamino)furan-2(5H)-one
Vzorec: C₁₁H₁₁NO₂
MW: 189,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03005353
CAS číslo: 38470-54-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR