Tisk…

6-Methylspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride

Dodavatel: Apollo Scientific
Varování

OR31009-1G OR31009-250MG
APOSOR31009-1GEA 38690 CZK
APOSOR31009-1G APOSOR31009-250MG
6-Methylspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
6-Methylspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride
Vzorec: C₁₃H₁₆N₂O₂·ClH
MW: 268,739 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29042695
CAS číslo: 1451154-33-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR