Tisk…

Methyl 3-bromobenzimidothioate hydroiodide salt

Dodavatel: Apollo Scientific
Varování

OR31007-1G OR31007-250MG
APOSOR31007-1GEA 787 CZK
APOSOR31007-1G APOSOR31007-250MG
Methyl 3-bromobenzimidothioate hydroiodide salt
Methyl 3-bromobenzimidothioate hydroiodide salt
Vzorec: C₈H₈BrNS·HI
Storage Temperature: Ambient
MDL Number: MFCD31812087
CAS číslo: 848564-76-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR