2-Methyl-2-[(propan-2-yl)amino]propanenitrile
Dodavatel: Apollo Scientific
OR310059-1G
OR310059-500MG
APOSOR310059-1GEA
9560
CZK
APOSOR310059-1G
APOSOR310059-500MG
2-Methyl-2-[(propan-2-yl)amino]propanenitrile
2-Methyl-2-[(propan-2-yl)amino]propanenitrile
Vzorec:
C₇H₁₄N₂ Storage Temperature: Ambient |
MDL Number:
MFCD06357358 CAS číslo: 16256-47-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...