4-Amino-3-methoxybenzamide 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR310052-1G
OR310052-500MG
APOSOR310052-1GEA
6880
CZK
APOSOR310052-1G
APOSOR310052-500MG
4-Amino-3-methoxybenzamide 95%
4-Amino-3-methoxybenzamide
Vzorec:
C₈H₁₀N₂O₂ MW: 166,177 g/mol Bod tání: 134…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18338357 CAS číslo: 211374-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...