7-Bromo-3-methylbenzofuran
Dodavatel: Apollo Scientific
|
Varování
|
OR31003-1G
OR31003-250MG
APOSOR31003-1GEA
14880
CZK
APOSOR31003-1G
APOSOR31003-250MG
7-Bromo-3-methylbenzofuran
7-Bromo-3-methylbenzofuran
Vzorec:
C₉H₇BrO MW: 211,055 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28405090 CAS číslo: 286837-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...