Tisk…

tert-Butyl 4-[(2-aminopyridin-3-yl)amino]piperidine-1-carboxylate

Dodavatel: Apollo Scientific

OR310002-1G OR310002-500MG
APOSOR310002-1GEA 9560 CZK
APOSOR310002-1G APOSOR310002-500MG
tert-Butyl 4-[(2-aminopyridin-3-yl)amino]piperidine-1-carboxylate
tert-Butyl 4-[(2-aminopyridin-3-yl)amino]piperidine-1-carboxylate
Vzorec: C₁₅H₂₄N₄O₂
MW: 292,377 g/mol
Bod tání: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD13184780
CAS číslo: 781649-86-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR