Tisk…

(2S,4R)-Methyl 4-hydroxypiperidine-2-carboxylate hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

OR30998-1G OR30998-250MG
APOSOR30998-1GEA 14390 CZK
APOSOR30998-1G APOSOR30998-250MG
(2S,4R)-Methyl 4-hydroxypiperidine-2-carboxylate hydrochloride 98%
(2S,4R)-Methyl 4-hydroxypiperidine-2-carboxylate hydrochloride
Vzorec: C₇H₁₃NO₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD11111603
CAS číslo: 175671-43-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR