2,7-Dichloroquinoline
Dodavatel: Apollo Scientific
OR309385-1G
OR309385-100MG
OR309385-500MG
OR309385-2.5G
OR309385-5G
APOSOR309385-1GEA
14880
CZK
APOSOR309385-1G
APOSOR309385-100MG
APOSOR309385-500MG
APOSOR309385-2.5G
APOSOR309385-5G
2,7-Dichloroquinoline
2,7-Dichloroquinoline
Vzorec:
C₉H₅Cl₂N MW: 198,049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00830402 CAS číslo: 613-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...