Tisk…

4-Bromoquinoline-3-carboxylic acid ethyl ester

Dodavatel: Apollo Scientific

Synonyms: Ethyl 4-bromoquinoline-3-carboxylate

OR309357-1G OR309357-250MG OR309357-5G
APOSOR309357-1GEA 2570 CZK
APOSOR309357-1G APOSOR309357-250MG APOSOR309357-5G
4-Bromoquinoline-3-carboxylic acid ethyl ester
4-Bromoquinoline-3-carboxylic acid ethyl ester
Vzorec: C₁₂H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD16987479
CAS číslo: 1242260-12-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR