Tisk…

3-Bromo-4-hydroxy-8-methoxyquinoline

Dodavatel: Apollo Scientific

OR309322-1G OR309322-5G
APOSOR309322-1GEA 8570 CZK
APOSOR309322-1G APOSOR309322-5G
3-Bromo-4-hydroxy-8-methoxyquinoline
3-Bromo-4-hydroxy-8-methoxyquinoline
Vzorec: C₁₀H₈BrNO₂
MW: 254,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13192861
CAS číslo: 1204811-42-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR