4-Hydroxy-7-methoxy-2-methylquinoline
Dodavatel: Apollo Scientific
OR309280-1G
OR309280-5G
APOSOR309280-1GEA
8570
CZK
APOSOR309280-1G
APOSOR309280-5G
4-Hydroxy-7-methoxy-2-methylquinoline
4-Hydroxy-7-methoxy-2-methylquinoline
Vzorec:
C₁₁H₁₁NO₂ MW: 189.211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00086643 CAS číslo: 103624-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...