Tisk…

6-Bromo-2-chloro-3-phenylquinoline

Dodavatel: Apollo Scientific

OR309271-1G OR309271-5G
APOSOR309271-1GEA 8100 CZK
APOSOR309271-1G APOSOR309271-5G
6-Bromo-2-chloro-3-phenylquinoline
6-Bromo-2-chloro-3-phenylquinoline
Vzorec: C₁₅H₉BrClN
MW: 318.596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997952
CAS číslo: 85274-48-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR