Tisk…

6-Bromo-4-hydroxy-2-methylquinoline

Dodavatel: Apollo Scientific

Synonyms: 6-Bromo-2-methylquinolin-4-ol

OR309248-5G
APOSOR309248-5GEA 3640 CZK
APOSOR309248-5G
6-Bromo-4-hydroxy-2-methylquinoline
6-Bromo-4-hydroxy-2-methylquinoline
Vzorec: C₁₀H₈BrNO
MW: 238,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00484433
CAS číslo: 103030-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR