6-Ethoxy-4-hydroxyquinoline
Dodavatel: Apollo Scientific
Synonyms:
6-Ethoxyquinolin-4-ol
OR309240-5G
APOSOR309240-5GEA
4930
CZK
APOSOR309240-5G
6-Ethoxy-4-hydroxyquinoline
6-Ethoxy-4-hydroxyquinoline
Vzorec:
C₁₁H₁₁NO₂ MW: 189,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02332227 CAS číslo: 303121-11-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...