Tisk…

8-Bromo-4-hydroxyquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific

OR309233-1G OR309233-5G
APOSOR309233-1GEA 7180 CZK
APOSOR309233-1G APOSOR309233-5G
8-Bromo-4-hydroxyquinoline-3-carboxylic acid
8-Bromo-4-hydroxyquinoline-3-carboxylic acid
Vzorec: C₁₀H₆BrNO₃
MW: 268,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179785
CAS číslo: 35973-17-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR