6-Chloro-4-hydroxyquinoline-3-carboxylic acid
Dodavatel: Apollo Scientific
OR309210-1G
OR309210-5G
APOSOR309210-1GEA
1160
CZK
APOSOR309210-1G
APOSOR309210-5G
6-Chloro-4-hydroxyquinoline-3-carboxylic acid
6-Chloro-4-hydroxyquinoline-3-carboxylic acid
Vzorec:
C₁₀H₆ClNO₃ MW: 223,613 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065198 CAS číslo: 35973-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...