Tisk…

6-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester

Dodavatel: Apollo Scientific

OR309206-1G OR309206-5G
APOSOR309206-1GEA 7180 CZK
APOSOR309206-1G APOSOR309206-5G
6-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
6-Chloro-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
Vzorec: C₁₄H₁₄ClNO₂
MW: 263,719 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09787816
CAS číslo: 948289-32-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR