Tisk…

5-Acetyl-1-methyl-1H-indazole 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Ethanoyl-1-methyl-1H-indazole, 1-(1-Methyl-1H-indazol-5-yl)ethan-1-one, 1-(1-Methyl-1H-indazol-5-yl)ethanone

OR30889-100MG OR30889-250MG OR30889-500MG
APOSOR30889-100MGEA 3310 CZK
APOSOR30889-100MG APOSOR30889-250MG APOSOR30889-500MG
5-Acetyl-1-methyl-1H-indazole 98%
5-Acetyl-1-methyl-1H-indazole
Vzorec: C₁₀H₁₀N₂O
MW: 174,199 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11869767
CAS číslo: 1159511-24-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR