Tisk…

7-(Aminomethyl)-2-methyl-2H-indazole

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Methyl-2H-indazol-7-yl)methylamine

OR30883-250MG OR30883-500MG
APOSOR30883-250MGEA 9330 CZK
APOSOR30883-250MG APOSOR30883-500MG
7-(Aminomethyl)-2-methyl-2H-indazole
7-(Aminomethyl)-2-methyl-2H-indazole
Vzorec: C₉H₁₁N₃
MW: 161,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869761
CAS číslo: 1144044-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR