5-(Aminomethyl)-1-methyl-1H-indazole 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR30872-250MG
OR30872-50MG
OR30872-100MG
APOSOR30872-250MGEA
9330
CZK
APOSOR30872-250MG
APOSOR30872-50MG
APOSOR30872-100MG
5-(Aminomethyl)-1-methyl-1H-indazole 95%
5-(Aminomethyl)-1-methyl-1H-indazole
Vzorec:
C₉H₁₁N₃ Storage Temperature: Ambient |
MDL Number:
MFCD12028599 CAS číslo: 267413-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...