Tisk…

1-Methyl-1H-indazole-5-carbonitrile 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Cyano-1-methyl-1H-indazole

OR30851-1G OR30851-250MG
APOSOR30851-1GEA 8570 CZK
APOSOR30851-1G APOSOR30851-250MG
1-Methyl-1H-indazole-5-carbonitrile 98%
1-Methyl-1H-indazole-5-carbonitrile
Vzorec: C₉H₇N₃
MW: 157,172 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12028575
CAS číslo: 189107-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR