(3S)-3-(Methoxymethyl)morpholine hydrochloride
Dodavatel: Apollo Scientific
Synonyms:
(S)-3-(Methoxymethyl)morpholine hydrochloride
OR308173-1G
OR308173-250MG
APOSOR308173-1GEA
4770
CZK
APOSOR308173-1G
APOSOR308173-250MG
(3S)-3-(Methoxymethyl)morpholine hydrochloride
(3S)-3-(Methoxymethyl)morpholine hydrochloride
Vzorec:
C₆H₁₃NO₂·ClH MW: 167.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09608107 CAS číslo: 218594-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...