Tisk…
Methyl morpholine-3-carboxylate hydrochloride
Dodavatel: Apollo Scientific
OR308165-1G OR308165-5G
APOSOR308165-1GEA 5100 CZK
APOSOR308165-1G APOSOR308165-5G
Methyl morpholine-3-carboxylate hydrochloride
Methyl morpholine-3-carboxylate hydrochloride
Vzorec: C₆H₁₁NO₃·ClH
MW: 181.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263285
CAS číslo: 1214686-81-3