Tisk…

tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate 97%

Dodavatel: Apollo Scientific

Synonyms: 1-Boc-(3S,4S)-3-amino-4-hydroxypyrrolidine

OR308146-1G OR308146-5G
APOSOR308146-1GEA 4570 CZK
APOSOR308146-1G APOSOR308146-5G
tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate 97%
tert-Butyl (3R,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate
Vzorec: C₉H₁₈N₂O₃
MW: 202,251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111251
CAS číslo: 190792-74-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR