Tisk…
tert-Butyl [4-(methylamino)butyl]carbamate
Dodavatel: Apollo Scientific
Synonyms: tert-Butyl (4-(methylamino)butyl)carbamate
OR308003-1G OR308003-5G
APOSOR308003-1GEA 4440 CZK
APOSOR308003-1G APOSOR308003-5G
tert-Butyl [4-(methylamino)butyl]carbamate
tert-Butyl [4-(methylamino)butyl]carbamate
Vzorec: C₁₀H₂₂N₂O₂
MW: 202.294 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06808587
CAS číslo: 874831-66-0