Tisk…

5-Chloro-2,8-dimethylquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific

OR307992-1G OR307992-5G
APOSOR307992-1GEA 8570 CZK
APOSOR307992-1G APOSOR307992-5G
5-Chloro-2,8-dimethylquinoline-3-carboxylic acid
5-Chloro-2,8-dimethylquinoline-3-carboxylic acid
Vzorec: C₁₂H₁₀ClNO₂
MW: 235,666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09787782
CAS číslo: 948294-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR