Tisk…

4-Hydroxy-8-methoxy-2-methylquinoline

Dodavatel: Apollo Scientific

OR307978-1G OR307978-5G
APOSOR307978-1GEA 2570 CZK
APOSOR307978-1G APOSOR307978-5G
4-Hydroxy-8-methoxy-2-methylquinoline
4-Hydroxy-8-methoxy-2-methylquinoline
Vzorec: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272321
CAS číslo: 15644-89-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR