4-Chloro-6-ethoxy-2-methylquinoline
Dodavatel: Apollo Scientific
OR307966-1G
OR307966-5G
APOSOR307966-1GEA
8830
CZK
APOSOR307966-1G
APOSOR307966-5G
4-Chloro-6-ethoxy-2-methylquinoline
4-Chloro-6-ethoxy-2-methylquinoline
Vzorec:
C₁₂H₁₂ClNO MW: 221,683 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02929376 CAS číslo: 66735-22-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...