4-Amino-8-methoxyquinoline
Dodavatel: Apollo Scientific
OR307959-1G
OR307959-5G
APOSOR307959-1GEA
9260
CZK
APOSOR307959-1G
APOSOR307959-5G
4-Amino-8-methoxyquinoline
4-Amino-8-methoxyquinoline
Vzorec:
C₁₀H₁₀N₂O MW: 174,199 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08063217 CAS číslo: 220844-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...