Tisk…

4-Amino-7-methoxyquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Amino-7-methoxylquinoline

OR307954-1G OR307954-250MG
APOSOR307954-1GEA 8900 CZK
APOSOR307954-1G APOSOR307954-250MG
4-Amino-7-methoxyquinoline
4-Amino-7-methoxyquinoline
Vzorec: C₁₀H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD08063213
CAS číslo: 103040-78-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR