2-Chloro-8-ethylquinoline-3-carbonitrile
Dodavatel: Apollo Scientific
OR307926-1G
OR307926-5G
APOSOR307926-1GEA
9260
CZK
APOSOR307926-1G
APOSOR307926-5G
2-Chloro-8-ethylquinoline-3-carbonitrile
2-Chloro-8-ethylquinoline-3-carbonitrile
Vzorec:
C₁₂H₉ClN₂ MW: 216,666 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09035781 CAS číslo: 880105-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...