2-Chloro-6,7-dimethylquinoline-3-carboxaldehyde
Dodavatel: Apollo Scientific
OR307915-1G
APOSOR307915-1GEA
14720
CZK
APOSOR307915-1G
2-Chloro-6,7-dimethylquinoline-3-carboxaldehyde
2-Chloro-6,7-dimethylquinoline-3-carboxaldehyde
Vzorec:
C₁₂H₁₀ClNO MW: 219,667 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02227046 CAS číslo: 94856-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...