Tisk…

2-Chloro-3-(2-chloroethyl)-7-methoxyquinoline

Dodavatel: Apollo Scientific

OR307898-1G OR307898-5G
APOSOR307898-1GEA 6680 CZK
APOSOR307898-1G APOSOR307898-5G
2-Chloro-3-(2-chloroethyl)-7-methoxyquinoline
2-Chloro-3-(2-chloroethyl)-7-methoxyquinoline
Vzorec: C₁₂H₁₁Cl₂NO
MW: 256,128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971676
CAS číslo: 73863-51-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR