Tisk…

2,8-Dimethylquinoline-3-carboxylic acid ethyl ester

Dodavatel: Apollo Scientific

OR307891-1G OR307891-5G
APOSOR307891-1GEA 8340 CZK
APOSOR307891-1G APOSOR307891-5G
2,8-Dimethylquinoline-3-carboxylic acid ethyl ester
2,8-Dimethylquinoline-3-carboxylic acid ethyl ester
Vzorec: C₁₄H₁₅NO₂
MW: 229,274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00510996
CAS číslo: 392734-40-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR