2,7-Dichloro-8-methylquinoline-3-carbonitrile
Dodavatel: Apollo Scientific
OR307888-1G
OR307888-5G
APOSOR307888-1GEA
8570
CZK
APOSOR307888-1G
APOSOR307888-5G
2,7-Dichloro-8-methylquinoline-3-carbonitrile
2,7-Dichloro-8-methylquinoline-3-carbonitrile
Vzorec:
C₁₁H₆Cl₂N₂ MW: 237,085 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09787545 CAS číslo: 948291-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...