Methyl 2-hydroxy-6-methylisonicotinate 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-hydroxy-6-methylpyridine-4-carboxylate, 2-Hydroxy-4-(methoxycarbonyl)-6-methylpyridine
OR30788-1G
OR30788-250MG
APOSOR30788-1GEA
827
CZK
APOSOR30788-1G
APOSOR30788-250MG
Methyl 2-hydroxy-6-methylisonicotinate 95+%
Methyl 2-hydroxy-6-methylisonicotinate
Vzorec:
C₈H₉NO₃ MW: 167.162 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13188667 CAS číslo: 98491-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...