6-Bromo-4-chloro-2-butylquinoline hydrochloride
Dodavatel: Apollo Scientific
OR307874-1G
OR307874-5G
APOSOR307874-1GEA
3380
CZK
APOSOR307874-1G
APOSOR307874-5G
6-Bromo-4-chloro-2-butylquinoline hydrochloride
6-Bromo-4-chloro-2-butylquinoline hydrochloride
Vzorec:
C₁₃H₁₃BrClN·ClH MW: 335,067 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10703348 CAS číslo: 1171071-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...