4-Chloro-2-propyl-6-quinolinecarbonitrile
Dodavatel: Apollo Scientific
OR307871-1G
OR307871-5G
APOSOR307871-1GEA
8570
CZK
APOSOR307871-1G
APOSOR307871-5G
4-Chloro-2-propyl-6-quinolinecarbonitrile
4-Chloro-2-propyl-6-quinolinecarbonitrile
Vzorec:
C₁₃H₁₁ClN₂ MW: 230,693 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09260432 CAS číslo: 930570-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...