6-Bromo-3-hydroxyquinoline 95%
Dodavatel: Apollo Scientific
Synonyms:
6-Bromoquinolin-3-ol
OR307869-1G
OR307869-250MG
APOSOR307869-1GEA
3940
CZK
APOSOR307869-1G
APOSOR307869-250MG
6-Bromo-3-hydroxyquinoline 95%
6-Bromo-3-hydroxyquinoline
Vzorec:
C₉H₆BrNO Storage Temperature: Ambient |
MDL Number:
MFCD09260430 CAS číslo: 552330-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...